Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001178133.1 | NP_001171604.1 | 3 | 559 | |
NM_016169.3 | NP_057253.2 | 3 | 559 | ||
XM_011539858.2 | XP_011538160.1 | 3 | 559 | ||
XM_011539860.2 | XP_011538162.1 | 3 | 559 | ||
XM_011539861.2 | XP_011538163.1 | 3 | 559 | ||
XM_011539863.2 | XP_011538165.1 | 3 | 294 | ||
XM_011539864.2 | XP_011538166.1 | 3 | 559 | ||
XM_017016323.1 | XP_016871812.1 | 3 | 789 | ||
GenBank mRNA | AF144231.1 | - | 3 | 514 | |
AF159447.1 | - | 3 | 514 | ||
AF172319.1 | - | 3 | 364 | ||
AF175770.1 | - | 3 | 534 | ||
AF222345.1 | - | 3 | 442 | ||
AK312319.1 | - | 3 | 549 | ||
AY358550.1 | - | 3 | 514 | ||
BC013291.2 | - | 3 | 535 | 3533158 | |
DA248208.1 | - | 3 | 559 |