Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001287489.1 | NP_001274418.1 | 30 | 3954 | |
NM_004802.3 | NP_004793.2 | 13 | 1752 | ||
NM_194248.2 | NP_919224.1 | 30 | 3954 | ||
NM_194322.2 | NP_919303.1 | 12 | 1906 | ||
NM_194323.2 | NP_919304.1 | 13 | 1752 | ||
XM_017005338.1 | XP_016860827.1 | 30 | 3767 | ||
GenBank mRNA | AF107403.1 | - | 13 | 1752 | |
AF183185.1 | - | 30 | 3954 | ||
AF183186.1 | - | 12 | 1906 | ||
AF183187.1 | - | 13 | 1752 | ||
AK296272.1 | - | 3 | 309 |