Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000355.3 | NP_000346.2 | 4 | 796 | |
NM_001184726.1 | NP_001171655.1 | 4 | 715 | ||
GenBank mRNA | AK225445.1 | - | 4 | 585 | |
AK313711.1 | - | 4 | 686 | ||
BC001176.1 | - | 4 | 674 | 3353271 | |
BC011239.1 | - | 4 | 594 | 4153362 | |
CR456591.1 | - | 4 | 586 | ||
L02647.1 | - | 4 | 644 | ||
L02648.1 | - | 4 | 600 | ||
M60396.1 | - | 4 | 584 |