Gene Transcripts
- Gene Symbol
- ANKRD20A5P
- Entrez Gene ID
- 440482
- Gene Name
- ankyrin repeat domain 20 family member A5, pseudogene
- Gene Aliases
- ANKRD20A5
- Chromosome Location
- Chr. 18: 14179097 - 14227050 on Build GRCh38
- UniGene ID
- Hs.632352
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC022023.1 |
- |
2 |
570 |
|
BC127726.1 |
- |
2 |
350 |
40127384 |
View Full Product Details