Gene Transcripts

Gene Symbol
AATK
Entrez Gene ID
9625
Gene Name
apoptosis associated tyrosine kinase
Gene Aliases
AATYK, AATYK1, LMR1, LMTK1, PPP1R77, p35BP
Chromosome Location
Chr. 17: 81117295 - 81166072 on Build GRCh38
UniGene ID
Hs.514575
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080395.2 NP_001073864.2 8 910
NM_004920.2 NP_004911.2 7 688
XM_006722193.2 XP_006722256.1 8 903
XM_006722194.2 XP_006722257.1 8 905
XM_006722195.2 XP_006722258.1 6 697
XM_006722196.2 XP_006722259.1 8 872
XM_011525509.2 XP_011523811.1 7 2339
XM_011525510.2 XP_011523812.1 7 715
XM_011525511.2 XP_011523813.1 3 568
XM_017025421.1 XP_016880910.1 7 702
XM_017025422.1 XP_016880911.1 7 861
GenBank mRNA AB014541.2 - 7 688
AK131395.1 - 6 701
AK131529.1 - 8 910

View Full Product Details