Gene Transcripts

Gene Symbol
GPR17
Entrez Gene ID
2840
Gene Name
G protein-coupled receptor 17
Gene Aliases
Chromosome Location
Chr. 2: 127645864 - 127652639 on Build GRCh38
UniGene ID
Hs.46453
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161415.1 NP_001154887.1 4 1032
NM_001161416.1 NP_001154888.1 3 967
NM_001161417.1 NP_001154889.1 2 738
NM_005291.2 NP_005282.1 3 803
XM_017003833.1 XP_016859322.1 2 1159
GenBank mRNA AK126849.1 - 1 1993
AK292725.1 - 2 608
AK313949.1 - 2 494
BC031653.1 - 2 417 5174146
BC039595.1 - 2 422
JF810891.1 - 1 337
Y12546.1 - 3 495
Z94154.1 - 2 431
Z94155.1 - 1 215

View Full Product Details