Gene Transcripts

Gene Symbol
Aicda
Entrez Gene ID
11628
Gene Name
activation-induced cytidine deaminase
Gene Aliases
Aid, Arp2
Chromosome Location
Chr. 6: 122553809 - 122564180 on Build GRCm38
UniGene ID
Mm.391503
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009645.2 NP_033775.1 3 396
GenBank mRNA AF132979.1 - 3 395
AK080144.1 - 2 395

View Full Product Details