Gene Transcripts

Gene Symbol
Hk1
Entrez Gene ID
15275
Gene Name
hexokinase 1
Gene Aliases
BB404130, Hk-1, Hk1-s, dea, mHk1-s
Chromosome Location
Chr. 10: 62268855 - 62379920 on Build GRCm38
UniGene ID
Mm.196605
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146100.1 NP_001139572.1 11 1947
NM_010438.3 NP_034568.2 15 1890
GenBank mRNA AK143630.1 - 11 1885
AK149749.1 - 11 1660
AK161466.1 - 5 859
AK167468.1 - 11 1703
AK185580.1 - 2 128
AK199185.1 - 2 128
BC027316.1 - 9 1042
BC072628.1 - 11 2068 6832742
J05277.1 - 11 1605
L16949.1 - 15 1869

View Full Product Details