Gene Transcripts

Gene Symbol
HOXC8
Entrez Gene ID
3224
Gene Name
homeobox C8
Gene Aliases
HOX3, HOX3A
Chromosome Location
Chr. 12: 54009106 - 54012763 on Build GRCh38
UniGene ID
Hs.664500
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022658.3 NP_073149.1 2 636
GenBank mRNA AK290959.1 - 2 627
BC053898.1 - 2 636 6171090
X99681.1 - 1 21

View Full Product Details