Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000861.3 | NP_000852.1 | 3 | 1388 | |
NM_001098211.1 | NP_001091681.1 | 2 | 1309 | ||
NM_001098212.1 | NP_001091682.1 | 2 | 1259 | ||
NM_001098213.1 | NP_001091683.1 | 2 | 1539 | ||
XM_011533652.1 | XP_011531954.1 | 4 | 1676 | ||
XM_011533653.2 | XP_011531955.1 | 2 | 1505 | ||
XM_017006283.1 | XP_016861772.1 | 2 | 1459 | ||
XM_017006284.1 | XP_016861773.1 | 2 | 1498 | ||
GenBank mRNA | AF026261.1 | - | 1 | 1197 | |
AK289412.1 | - | 2 | 1539 | ||
AK292157.1 | - | 2 | 1256 | ||
AK312879.1 | - | 2 | 1257 | ||
AY136743.1 | - | 1 | 1197 | ||
BC060802.1 | - | 3 | 1388 | 30340368 | |
D28481.1 | - | 1 | 1200 | ||
Z34897.1 | - | 1 | 1197 |