Gene Transcripts

Gene Symbol
Ptch1
Entrez Gene ID
19206
Gene Name
patched homolog 1
Gene Aliases
A230106A15Rik, Ptc, Ptc1, Ptch, mes
Chromosome Location
Chr. 13: 63508328 - 63573616 on Build GRCm38
UniGene ID
Mm.228798
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008957.2 NP_032983.1 15 2414
GenBank mRNA AK147626.1 - 16 2561
U46155.1 - 15 2414

View Full Product Details