Gene Transcripts

Gene Symbol
Exoc4
Entrez Gene ID
20336
Gene Name
exocyst complex component 4
Gene Aliases
C78892, Sec8, Sec8l1
Chromosome Location
Chr. 6: 33249090 - 33972943 on Build GRCm38
UniGene ID
Mm.265512
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009148.3 NP_033174.2 2 148
GenBank mRNA AF022962.1 - 2 174
AK010695.1 - 2 163
AK014499.1 - 2 171
AK036507.1 - 2 160
AK038125.1 - 2 171
AK039983.1 - 2 171
AK086603.1 - 2 167
AK173235.1 - 2 151
BC034644.1 - 2 148 5355276

View Full Product Details