Gene Transcripts

Gene Symbol
Tom1
Entrez Gene ID
21968
Gene Name
target of myb1 homolog (chicken)
Gene Aliases
Chromosome Location
Chr. 8: 75033686 - 75070121 on Build GRCm38
UniGene ID
Mm.1967
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136259.1 NP_001129731.1 10 1093
NM_011622.3 NP_035752.1 10 1093
GenBank mRNA AJ006972.1 - 10 1070
AK028398.1 - 10 1034
AK051245.1 - 10 1075
AK150143.1 - 10 1043
AK160403.1 - 10 1039
AK182481.1 - 1 17
BC021633.1 - 10 1056 4208226
BC093520.1 - 10 1021 2866013
BC157905.1 - 10 1054 9088106
BC158042.1 - 10 1054 9007343

View Full Product Details