Gene Transcripts

Gene Symbol
Wnt5b
Entrez Gene ID
22419
Gene Name
wingless-type MMTV integration site family, member 5B
Gene Aliases
AW545702, Wnt-5b
Chromosome Location
Chr. 6: 119432531 - 119544824 on Build GRCm38
UniGene ID
Mm.321818
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271757.1 NP_001258686.1 4 772
NM_001271758.1 NP_001258687.1 4 715
NM_009525.3 NP_033551.2 4 969
GenBank mRNA AK013218.1 - 4 940
AK049558.1 - 4 739
BC010775.1 - 4 715 4211437
BC051406.1 - 4 900 6438917
BU520338.1 - 2 381
M89799.1 - 3 584

View Full Product Details