Gene Transcripts
- Gene Symbol
- Golga5
- Entrez Gene ID
- 27277
- Gene Name
- golgi autoantigen, golgin subfamily a, 5
- Gene Aliases
- Ret-II
- Chromosome Location
- Chr. 12: 102469134 - 102497911 on Build GRCm38
- UniGene ID
- Mm.273370
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199004.1 |
NP_001185933.1
|
4, 5 |
1240 |
|
NM_013747.4 |
NP_038775.1
|
4, 5 |
1082 |
|
GenBank mRNA |
AB016784.1 |
- |
4, 5 |
1241 |
|
AF026274.1 |
- |
4, 5 |
1072 |
|
AK007592.1 |
- |
4, 5 |
1084 |
|
AK138455.1 |
- |
4, 5 |
1241 |
|
AK152533.1 |
- |
4, 5 |
1090 |
|
AK153010.1 |
- |
4, 5 |
1090 |
|
AK168765.1 |
- |
4, 5 |
1086 |
|
BC016883.1 |
- |
4, 5 |
1810 |
4235952 |
BC086782.1 |
- |
4, 5 |
1082 |
6813878 |
View Full Product Details