Gene Transcripts

Gene Symbol
Timm8b
Entrez Gene ID
30057
Gene Name
translocase of inner mitochondrial membrane 8B
Gene Aliases
Tim8b
Chromosome Location
Chr. 9: 50603901 - 50605320 on Build GRCm38
UniGene ID
Mm.335149
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013897.2 NP_038925.1 1, 2 147
GenBank mRNA AA204614.1 - 1, 2 136
AF196314.1 - 136
AK003382.1 - 1, 2 106
AK004190.1 - 136
AK012925.1 - 1, 2 114
AK018718.1 - 1, 2 114
AK019422.1 - 1, 2 122
AK169258.1 - 1, 2 126
BC029239.1 - 1, 2 100 4237232
BY100901.1 - 1, 2 151

View Full Product Details