Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000420.2 | NP_000411.1 | 17 | 2067 | |
XM_005249993.2 | XP_005250050.1 | 17 | 1918 | ||
XM_005249994.3 | XP_005250051.1 | 11 | 1238 | ||
GenBank mRNA | AK314831.1 | - | 17 | 1968 | |
BC003135.1 | - | 17 | 2067 | 2906155 | |
BC050639.1 | - | 17 | 1962 | 5742350 | |
FN998902.1 | - | 16 | 1857 | ||
HE572750.1 | - | 16 | 1857 | ||
KC922434.1 | - | 2 | 202 | ||
M64934.2 | - | 17 | 1977 |