Gene Transcripts
- Gene Symbol
- Ccnl2
- Entrez Gene ID
- 56036
- Gene Name
- cyclin L2
- Gene Aliases
- 1700010A01Rik, 1810019L15Rik, 2010319M22Rik, AA409936, AW261738, Ania-6b, HCLA-150, Pcee, SB138
- Chromosome Location
- Chr. 4: 155812084 - 155824543 on Build GRCm38
- UniGene ID
- Mm.23492
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207678.1 |
NP_997561.1
|
4, 5 |
626 |
|
GenBank mRNA |
AB041605.1 |
- |
4, 5 |
603 |
|
AF211859.1 |
- |
4, 5 |
587 |
|
AK007552.1 |
- |
4, 5 |
623 |
|
AK008585.1 |
- |
4, 5 |
415 |
|
AK044204.1 |
- |
4, 5 |
563 |
|
AK048244.1 |
- |
4, 5 |
616 |
|
AK156037.1 |
- |
4, 5 |
609 |
|
AK169857.1 |
- |
4, 5 |
602 |
|
AY337018.1 |
- |
4, 5 |
609 |
|
BC023747.1 |
- |
4, 5 |
601 |
|
BC132295.1 |
- |
4, 5 |
618 |
40130572 |
CD348864.1 |
- |
4, 5 |
626 |
|
View Full Product Details