Gene Transcripts

Gene Symbol
Sft2d3
Entrez Gene ID
67158
Gene Name
SFT2 domain containing 3
Gene Aliases
2610206C24Rik, AI847965, AW046173
Chromosome Location
Chr. 18: 31909094 - 31911903 on Build GRCm38
UniGene ID
Mm.31537
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026006.1 NP_080282.1 1 2019
GenBank mRNA AK011898.1 - 1 1394

View Full Product Details