Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001099855.1 | NP_001093325.1 | 11 | 1245 | |
NM_001171876.1 | NP_001165347.1 | 11 | 1245 | ||
NM_001171877.1 | NP_001165348.1 | 7 | 1110 | ||
NM_001171878.1 | NP_001165349.1 | 8 | 1227 | ||
NM_001171879.1 | NP_001165350.1 | 8 | 1227 | ||
NM_005369.4 | NP_005360.3 | 8 | 1227 | ||
XM_005262413.4 | XP_005262470.1 | 12 | 1393 | ||
XM_011531339.2 | XP_011529641.1 | 12 | 1452 | ||
XM_017029529.1 | XP_016885018.1 | 11 | 1214 | ||
XM_017029530.1 | XP_016885019.1 | 11 | 1214 | ||
XM_017029531.1 | XP_016885020.1 | 11 | 1147 | ||
XM_017029532.1 | XP_016885021.1 | 8 | 1304 | ||
GenBank mRNA | AB085901.1 | - | 8 | 904 | |
AB085902.1 | - | 8 | 904 | ||
AK296488.1 | - | 7 | 1110 | ||
AK296529.1 | - | 11 | 1345 | ||
AK302957.1 | - | 12 | 1193 | ||
AK313900.1 | - | 8 | 1087 | ||
AK316416.1 | - | 11 | 1233 | ||
AL117234.1 | - | 10 | 1128 | ||
X12556.1 | - | 8 | 1078 |