Gene Transcripts

Gene Symbol
Tspan11
Entrez Gene ID
68498
Gene Name
tetraspanin 11
Gene Aliases
1110014F12Rik, AU067806
Chromosome Location
Chr. 6: 127887622 - 127953031 on Build GRCm38
UniGene ID
Mm.174368
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026743.3 NP_081019.1 8 1552
GenBank mRNA AK028808.1 - 8 1443
AK053788.1 - 8 1539
AK090022.1 - 8 1553

View Full Product Details