Gene Transcripts

Gene Symbol
Ing2
Entrez Gene ID
69260
Gene Name
inhibitor of growth family, member 2
Gene Aliases
2810011M06Rik, Ing1l, Ing2b, P33ING2
Chromosome Location
Chr. 8: 47667178 - 47675609 on Build GRCm38
UniGene ID
Mm.430733
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023503.3 NP_075992.2 2 1215
GenBank mRNA AB433625.1 - 2 718
AF078834.1 - 2 743
AK012716.1 - 2 1212
AK048800.1 - 2 1211
AK083144.1 - 2 1214
BC050003.1 - 2 1160
BC096433.1 - 2 1195 6310342
BC138229.1 - 2 946 8861248

View Full Product Details