Gene Transcripts

Gene Symbol
Tmem52
Entrez Gene ID
69671
Gene Name
transmembrane protein 52
Gene Aliases
2310043I08Rik
Chromosome Location
Chr. 4: 155469114 - 155470858 on Build GRCm38
UniGene ID
Mm.45401
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253856.1 NP_001240785.1 5 775
NM_001253858.1 NP_001240787.1 4 733
NM_027161.2 NP_081437.1 5 793
GenBank mRNA AK009779.1 - 5 756
BC144992.1 - 5 792 9053540
BC150685.1 - 5 773 9087596

View Full Product Details