Gene Transcripts

Gene Symbol
Cdk3-ps
Entrez Gene ID
69681
Gene Name
cyclin-dependent kinase 3, pseudogene
Gene Aliases
2310050O10Rik, Cdk3
Chromosome Location
Chr. 11: 116216002 - 116220287 on Build GRCm38
UniGene ID
Mm.33677
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK009918.1 - 5 888
AK166387.1 - 2 375
BC116895.1 - 5 798
BC119034.1 - 5 798

View Full Product Details