Gene Transcripts

Gene Symbol
Rnaseh2a
Entrez Gene ID
69724
Gene Name
ribonuclease H2, large subunit
Gene Aliases
2400006P09Rik, RNASEHI, RNHIA, RNHL
Chromosome Location
Chr. 8: 84956610 - 84969375 on Build GRCm38
UniGene ID
Mm.182470
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027187.3 NP_081463.1 3 266
GenBank mRNA AK010292.1 - 3 256
AK146518.1 - 3 267
BC029252.1 - 3 296 3666709
BC038158.1 - 3 340 1493985

View Full Product Details