Gene Transcripts

Gene Symbol
Cysltr2
Entrez Gene ID
70086
Gene Name
cysteinyl leukotriene receptor 2
Gene Aliases
2300001H05Rik, CYSLT2R, Cltr2, Cyslt2
Chromosome Location
Chr. 14: 73026075 - 73049114 on Build GRCm38
UniGene ID
Mm.158324
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162412.1 NP_001155884.1 5 1241
NM_133720.3 NP_598481.2 6 1298
GenBank mRNA AF331658.1 - 6 1257
AK008997.1 - 5 1236

View Full Product Details