Gene Transcripts

Gene Symbol
Asun
Entrez Gene ID
71177
Gene Name
asunder, spermatogenesis regulator
Gene Aliases
4933424B01Rik, Spata30
Chromosome Location
Chr. 6: 146549632 - 146577835 on Build GRCm38
UniGene ID
Mm.271774
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138757.2 NP_620096.2 2 723
GenBank mRNA BC026851.1 - 2 652
BC027229.1 - 2 308 2615769
BY741532.1 - 2 723
CF167981.1 - 2 319

View Full Product Details