Gene Transcripts

Gene Symbol
5430416O09Rik
Entrez Gene ID
71406
Gene Name
RIKEN cDNA 5430416O09 gene
Gene Aliases
AI481365
Chromosome Location
Chr. 4: 43730034 - 43734534 on Build GRCm38
UniGene ID
Mm.33063
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK017317.1 - 2 1368
AK030334.1 - 2 1386
BC064043.1 - 2 1353
BC147344.1 - 1 917 9056418
BC147345.1 - 1 917 9056411

View Full Product Details