Gene Transcripts

Gene Symbol
Cwf19l1
Entrez Gene ID
72502
Gene Name
CWF19-like 1, cell cycle control (S. pombe)
Gene Aliases
2610528C06Rik, AI854304, AV336991
Chromosome Location
Chr. 19: 44108637 - 44135878 on Build GRCm38
UniGene ID
Mm.276109
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081077.1 NP_001074546.1 10 1094
GenBank mRNA AK087867.1 - 4 367
AK088543.1 - 12 2336
AK139654.1 - 10 1095
BC037640.1 - 10 1072 5096937

View Full Product Details