Gene Transcripts

Gene Symbol
Usp13
Entrez Gene ID
72607
Gene Name
ubiquitin specific peptidase 13 (isopeptidase T-3)
Gene Aliases
2700071E21Rik, AI848077, ISOT3, IsoT-3
Chromosome Location
Chr. 3: 32817019 - 32938075 on Build GRCm38
UniGene ID
Mm.316153
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013024.2 NP_001013042.1 4 459
GenBank mRNA AK135141.1 - 4 511
BC090999.1 - 4 459 6754549

View Full Product Details