Gene Transcripts

Gene Symbol
Cltb
Entrez Gene ID
74325
Gene Name
clathrin, light polypeptide (Lcb)
Gene Aliases
2310046E19Rik
Chromosome Location
Chr. 13: 54592939 - 54611397 on Build GRCm38
UniGene ID
Mm.290026
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028870.3 NP_083146.1 4 447
GenBank mRNA AK009844.1 - 2 108
AK029382.1 - 4 448
AK045989.1 - 4 468
AK146816.1 - 520
AK159480.1 - 4 497
AK167531.1 - 520
AK213693.1 - 2 130
BC037505.1 - 1 82
BC070404.1 - 4 449 5699653
BC128349.1 - 1 73

View Full Product Details