Gene Transcripts

Gene Symbol
Nipal3
Entrez Gene ID
74552
Gene Name
NIPA-like domain containing 3
Gene Aliases
9130020G22Rik, AI480660, Npal3
Chromosome Location
Chr. 4: 135447136 - 135494806 on Build GRCm38
UniGene ID
Mm.26548
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028995.3 NP_083271.1 11 1296
GenBank mRNA AK018640.1 - 11 1273
AK049696.1 - 11 1331
AK053937.1 - 1300
AK082376.1 - 1298
AK087811.1 - 11 1299
AK138843.1 - 11 1297
AK148206.1 - 11 1318
BC057168.1 - 11 1236 4973200

View Full Product Details