Gene Transcripts

Gene Symbol
Mdga1
Entrez Gene ID
74762
Gene Name
MAM domain containing glycosylphosphatidylinositol anchor 1
Gene Aliases
1200011I03Rik, GPIM, Mamdc3
Chromosome Location
Chr. 17: 29827956 - 29888429 on Build GRCm38
UniGene ID
Mm.40901
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081160.1 NP_001074629.1 15 3113
GenBank mRNA AK004706.1 - 8 1123
DQ788983.1 - 3542

View Full Product Details