Gene Transcripts

Gene Symbol
Fam35a
Entrez Gene ID
75698
Gene Name
family with sequence similarity 35, member A
Gene Aliases
3110001K24Rik
Chromosome Location
Chr. 14: 34236948 - 34310503 on Build GRCm38
UniGene ID
Mm.45406
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029389.2 NP_083665.2 4 973
GenBank mRNA AK149442.1 - 4 837
AK162134.1 - 2 784
BC144822.1 - 1 481

View Full Product Details