Gene Transcripts

Gene Symbol
Adal
Entrez Gene ID
75894
Gene Name
adenosine deaminase-like
Gene Aliases
4930578F03Rik
Chromosome Location
Chr. 2: 121140428 - 121156680 on Build GRCm38
UniGene ID
Mm.384805
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290811.1 NP_001277740.1 10 1160
NM_001290812.1 NP_001277741.1 9 1054
NM_029475.2 NP_083751.1 10 1052
GenBank mRNA AK039677.1 - 9 1045
AK049697.1 - 10 1153
BC050879.1 - 10 1004 5687835
BC052048.1 - 10 1004 5687835

View Full Product Details