Gene Transcripts

Gene Symbol
Memo1
Entrez Gene ID
76890
Gene Name
mediator of cell motility 1
Gene Aliases
0610016J10Rik, D930048L02Rik
Chromosome Location
Chr. 17: 74198398 - 74295544 on Build GRCm38
UniGene ID
Mm.311874
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133771.2 NP_598532.1 7 754
GenBank mRNA AF363447.1 - 8 816
AK002639.1 - 2 159
AK159560.1 - 7 733
BC013819.1 - 7 771 3489363

View Full Product Details