Gene Transcripts

Gene Symbol
Rsph6a
Entrez Gene ID
83434
Gene Name
radial spoke head 6 homolog A (Chlamydomonas)
Gene Aliases
RSP4, Rshl1
Chromosome Location
Chr. 7: 19054678 - 19074447 on Build GRCm38
UniGene ID
Mm.84435
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159671.1 NP_001153143.1 1 483
NM_031255.2 NP_112545.2 1 483
GenBank mRNA AF329191.1 - 1 471
AF329192.1 - 1 476
AK029699.1 - 1 483
BC137992.1 - 1 476 8861011

View Full Product Details