Gene Transcripts

Gene Symbol
Pi4k2a
Entrez Gene ID
84095
Gene Name
phosphatidylinositol 4-kinase type 2 alpha
Gene Aliases
Pi4k2
Chromosome Location
Chr. 19: 42090146 - 42122218 on Build GRCm38
UniGene ID
Mm.117037
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145501.2 NP_663476.1 5 1010
GenBank mRNA AK030725.1 - 5 975
AK185438.1 - 1 31
BC089309.1 - 5 951
BC110363.1 - 5 972 5346917

View Full Product Details