Gene Transcripts
- Gene Symbol
- Hivep1
- Entrez Gene ID
- 110521
- Gene Name
- human immunodeficiency virus type I enhancer binding protein 1
- Gene Aliases
- Cryabp1
- Chromosome Location
- Chr. 13: 42051969 - 42185030 on Build GRCm38
- UniGene ID
- Mm.4808
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_007772.2 |
NP_031798.2
|
4 |
2543 |
|
View Full Product Details