Gene Transcripts

Gene Symbol
Slc7a6
Entrez Gene ID
330836
Gene Name
solute carrier family 7 (cationic amino acid transporter, y+ system), member 6
Gene Aliases
AI643885, LAT-2, LAT3
Chromosome Location
Chr. 8: 106168830 - 106198708 on Build GRCm38
UniGene ID
Mm.46749
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178798.3 NP_848913.1 10 3099
GenBank mRNA AK049478.1 - 10 3099
BC042898.1 - 7 3631

View Full Product Details