Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006446.4 | NP_006437.3 | 8 | 849 | |
GenBank mRNA | AB026257.1 | - | 8 | 844 | |
AF060500.1 | - | 7 | 745 | ||
AF205071.1 | - | 8 | 879 | ||
AJ132573.1 | - | 7 | 808 | ||
AK312970.1 | - | 8 | 831 | ||
BC005243.2 | - | 7 | 807 | ||
BC020696.1 | - | 8 | 850 | ||
BC054878.1 | - | 8 | 837 | ||
BC070172.1 | - | 8 | 847 | ||
BC105596.1 | - | 8 | 837 | ||
BC114376.1 | - | 8 | 844 | 40037364 |