Gene Transcripts
- Gene Symbol
- PGLYRP4
- Entrez Gene ID
- 57115
- Gene Name
- peptidoglycan recognition protein 4
- Gene Aliases
- PGLYRPIbeta, PGRP-Ibeta, PGRPIB, SBBI67
- Chromosome Location
- Chr. 1: 153327410 - 153348844 on Build GRCh38
- UniGene ID
- Hs.58356
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020393.3 |
NP_065126.2
|
8, 9 |
1311 |
|
XM_011509789.2 |
XP_011508091.1
|
8, 9 |
1323 |
|
XM_011509790.1 |
XP_011508092.1
|
8, 9 |
1311 |
|
XM_011509791.2 |
XP_011508093.1
|
8, 9 |
1302 |
|
XM_011509793.2 |
XP_011508095.1
|
5, 6 |
999 |
|
GenBank mRNA |
AF242518.1 |
- |
8, 9 |
1024 |
|
AK292203.1 |
- |
8, 9 |
1314 |
|
AY035377.1 |
- |
8, 9 |
1016 |
|
BC107157.1 |
- |
7, 8 |
998 |
40032748 |
BC107158.1 |
- |
7, 8 |
998 |
40032750 |
BC142636.1 |
- |
8, 9 |
1286 |
40147772 |
View Full Product Details