Gene Transcripts

Gene Symbol
ST6GAL2
Entrez Gene ID
84620
Gene Name
ST6 beta-galactoside alpha-2,6-sialyltransferase 2
Gene Aliases
SIAT2, ST6GalII
Chromosome Location
Chr. 2: 106801600 - 106887410 on Build GRCh38
UniGene ID
Hs.98265
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142351.1 NP_001135823.1 4, 5 1267
NM_001142352.1 NP_001135824.1 4, 5 1337
NM_001322362.1 NP_001309291.1 4, 5 1316
NM_032528.2 NP_115917.1 4, 5 1337
NR_136313.1 3, 4 352
XM_006712802.1 XP_006712865.1 4, 5 1352
XM_011511999.2 XP_011510301.1 4, 5 1449
XM_011512000.2 XP_011510302.1 4, 5 1534
XM_011512001.2 XP_011510303.1 4, 5 1534
XM_017005110.1 XP_016860599.1 4, 5 1288
GenBank mRNA AB058780.1 - 4, 5 1267
AB059555.1 - 4, 5 1323
AJ512141.1 - 2, 3 208
AK307784.1 - 4, 5 1337
BC008680.1 - 4, 5 1323 3856171

View Full Product Details