Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025003.3 | NP_079279.3 | 12 | 1698 | |
XM_011538754.2 | XP_011537056.1 | 12 | 1751 | ||
XM_017019979.1 | XP_016875468.1 | 5 | 767 | ||
GenBank mRNA | AF488804.1 | - | 12 | 1698 | |
AJ515153.3 | - | 12 | 1698 | ||
AJ515154.2 | - | 12 | 1698 |