Gene Transcripts

Gene Symbol
PGLYRP4
Entrez Gene ID
57115
Gene Name
peptidoglycan recognition protein 4
Gene Aliases
PGLYRPIbeta, PGRP-Ibeta, PGRPIB, SBBI67
Chromosome Location
Chr. 1: 153327410 - 153348844 on Build GRCh38
UniGene ID
Hs.58356
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020393.3 NP_065126.2 7 1065
XM_011509789.2 XP_011508091.1 7 1077
XM_011509790.1 XP_011508092.1 7 1065
XM_011509791.2 XP_011508093.1 7 1056
XM_011509792.1 XP_011508094.1 7 1065
XM_011509793.2 XP_011508095.1 4 753
GenBank mRNA AF242518.1 - 7 778
AK292203.1 - 7 1068
AY035377.1 - 7 770
BC107157.1 - 6 752 40032748
BC107158.1 - 6 752 40032750
BC142636.1 - 7 1040 40147772

View Full Product Details