Gene Transcripts

Gene Symbol
ADPRHL1
Entrez Gene ID
113622
Gene Name
ADP-ribosylhydrolase like 1
Gene Aliases
ARH2
Chromosome Location
Chr. 13: 113421940 - 113453524 on Build GRCh38
UniGene ID
Hs.98669
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138430.4 NP_612439.2 2 399
NM_199162.2 NP_954631.1 2 242
GenBank mRNA AJ313429.1 - 2 399
BC009755.2 - 2 242 3838063

View Full Product Details