Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001003801.1 | NP_001003801.1 | 4 | 498 | |
NM_001003802.1 | NP_001003802.1 | 5 | 487 | ||
NM_003078.3 | NP_003069.2 | 5 | 526 | ||
XM_011516521.2 | XP_011514823.1 | 4 | 605 | ||
XM_017012556.1 | XP_016868045.1 | 4 | 325 | ||
GenBank mRNA | AK127595.1 | - | 5 | 1034 | |
AK297876.1 | - | 4 | 520 | ||
AK309500.1 | - | 4 | 483 | ||
AK316173.1 | - | 4 | 470 | ||
AY450430.1 | - | 4 | 802 | ||
AY450431.1 | - | 5 | 487 | ||
BC002628.2 | - | 5 | 487 | 3608512 | |
BC110350.1 | - | 4 | 417 | 5212363 | |
BX648385.1 | - | 4 | 264 | ||
U66619.1 | - | 5 | 535 |