Gene Transcripts

Gene Symbol
BIN3
Entrez Gene ID
55909
Gene Name
bridging integrator 3
Gene Aliases
Chromosome Location
Chr. 8: 22620418 - 22669148 on Build GRCh38
UniGene ID
Hs.433722
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018688.4 NP_061158.1 4, 5 261
XM_011544587.1 XP_011542889.1 4, 5 261
GenBank mRNA AF271732.1 - 4, 5 224
AK023980.1 - 4, 5 208
AK222470.1 - 4, 5 233
BC001223.1 - 3, 4 145 3534031
BC009824.2 - 4, 5 195 3953604

View Full Product Details