Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021254.2 | NP_067077.1 | 2 | 630 | |
NR_036552.1 | 2 | 625 | |||
GenBank mRNA | AF282851.1 | - | 2 | 378 | |
AK000474.1 | - | 2 | 625 | ||
AK055328.1 | - | 3 | 567 | ||
AK097566.1 | - | 1531 | |||
AK098577.1 | - | 2 | 630 | ||
AK223318.1 | - | 2 | 608 | ||
AK225697.1 | - | 2 | 608 | ||
BC000709.2 | - | 2 | 383 | 3504049 | |
FJ649644.1 | - | 2 | 1558 |