Gene Transcripts

Gene Symbol
TEKT2
Entrez Gene ID
27285
Gene Name
tektin 2
Gene Aliases
TEKTB1, TEKTIN-T, h-tektin-t
Chromosome Location
Chr. 1: 36084075 - 36088275 on Build GRCh38
UniGene ID
Hs.127111
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014466.2 NP_055281.2 6 858
XM_005270753.2 XP_005270810.1 6 1174
XM_011541258.2 XP_011539560.1 6 870
XM_017001055.1 XP_016856544.1 6 830
GenBank mRNA AB033823.1 - 5 790
AF054910.1 - 6 858
AK313610.1 - 6 805
BC035620.1 - 6 808 5172213

View Full Product Details